missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5415103

 View more versions of this product

Catalog No. AAH5415103



Specifications

Specifications

2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester
1g
C14H21BN2O3
2-ethylaminocarbonyl pyridine-5-boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)NCC
276.143
276.14
52°C to 54°C
1006876-28-3
MFCD13191367
TVMRHMZREIYXSR-UHFFFAOYSA-N
N-ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
59868917
96%

Chemical Identifiers

1006876-28-3
276.143
TVMRHMZREIYXSR-UHFFFAOYSA-N
59868917
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)NCC
C14H21BN2O3
MFCD13191367
2-ethylaminocarbonyl pyridine-5-boronic acid pinacol ester
N-ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents