Learn More
2-Ethylbutyryl chloride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2377422
Description
2-Ethylbutyryl chloride was used in syntheses of bicyclic thiophene derivatives. It is used as an organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Ethylbutyryl chloride was used in syntheses of bicyclic thiophene derivatives. It is used as an organic intermediate.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, amines, alcohols, active metals, water/ moisture.
Specifications
2-Ethylbutyryl chloride | |
0.98 | |
33°C (91°F) | |
C6H11ClO | |
MFCD00018812 | |
UN3094 | |
2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride | |
SMUKODJVMQOSAB-UHFFFAOYSA-N | |
2-ethylbutanoyl chloride | |
75954 | |
99% |
2736-40-5 | |
137°C to 139°C | |
Acrid | |
1.425 | |
100 g | |
Moisture sensitive | |
Reacts with water. | |
CCC(CC)C(Cl)=O | |
134.60 | |
134.61 |
Chemical Identifiers
2736-40-5 | |
134.60 | |
SMUKODJVMQOSAB-UHFFFAOYSA-N | |
75954 | |
CCC(CC)C(Cl)=O |
C6H11ClO | |
MFCD00018812 | |
2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride | |
2-ethylbutanoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber : 220-359-9
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only