Please login to your online account to display your discounted pricing

2-Ethylbutyric Acid 99%, ACROS Organics™

$18.60 - $18.60

Chemical Identifiers

Chemical Name or Material 2-Ethylbutyric acid
Name Note 99%
CAS 88-09-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H12O2
Linear Formula (C2H5)2CHCO2H
Formula Weight 116.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC149410050

  SDS
thermo fisher scientific
149410050
Glass bottle 5g Each for $18.60
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 116.16
Quantity 5g
Color Clear colorless to light yellow
EINECS Number 201-796-4
TSCA TSCA
RTECS Number ET1400000
Density 0.9200g/mL
Beilstein 02, 333
Merck Index 15, 3121
CAS Max % 100.0
Chemical Name or Material 2-Ethylbutyric acid
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4120 to 1.4140 (20°C, 589nm)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H12O2
Linear Formula (C2H5)2CHCO2H
MDL Number MFCD00002670
CAS 88-09-5
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.