Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethylbutyraldehyde, 98%

Chemical Identifiers

Chemical Name or Material 2-Ethylbutyraldehyde
CAS 97-96-1
Molecular Formula C6H12O
Formula Weight 100.16g/mol
MDL Number MFCD00006985
UN Number UN1178
Beilstein 1209330
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB23465AK

 
alfa aesar™
B23465-AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 116° to 118°C
Quantity 250mL
Flash Point 8°C (46°F)
Odor Like Aldehyde
Chemical Name or Material 2-Ethylbutyraldehyde
CAS 97-96-1
Assay 98%
Molecular Formula C6H12O
Formula Weight 100.16g/mol
MDL Number MFCD00006985
UN Number UN1178
Beilstein 1209330
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H319-H335
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-ETHYLBUTYRALDEHYDE
RTECS Number ES2625000
TSCA TSCA

Applications
2-Ethylbutyraldehyde is used in the preparation of aldoximes using aqueous hydroxylamine. It is used as pharmaceutical intermediates.

Solubility
Soluble in ether and alcohols. Slightly soluble in water.

Notes
Air Sensitive. Store under dry inert gas. Store away from air.

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H319-H335
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.