Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethylbutyl acetate, 97%

Chemical Identifiers

Chemical Name or Material 2-Ethylbutyl acetate
CAS 10031-87-5
Molecular Formula CH3CO2CH2CH(CH2CH3)2
Formula Weight 144.22
MDL Number MFCD00009343
UN Number UN1177
Beilstein 1746228
Synonym Acetic acid 2-ethylbutyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0551614

  SDS
alfa aesar™
L0551614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0551622

  SDS
alfa aesar™
L0551622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.22
CAS 10031-87-5
Chemical Name or Material 2-Ethylbutyl acetate
Health Hazard 1 Warning
MDL Number MFCD00009343
TSCA TSCA
Assay 97%
UN Number UN1177
Molecular Formula CH3CO2CH2CH(CH2CH3)2
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 2-ETHYLBUTYL ACETATE
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 1746228
Boiling Point 159° to 160°C
Flash Point 52°C (125°F)
Quantity 25g
Synonym Acetic acid 2-ethylbutyl ester
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.