missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylbenzenethiol, TCI America™

Manufacturer:  TCI America E070225G

 View more versions of this product

Catalog No. E070225G



Specifications

Specifications

2-Ethylbenzenethiol
Yellow
4500-58-7
MFCD00010022
ABROBCBIIWHVNS-UHFFFAOYSA-N
2-ethylbenzenethiol
3734338
Clear Liquid at 20°C
205°C
25g
C8H10S
2-ethylthiophenol, benzenethiol, 2-ethyl, unii-62gbu45i5p, 2-ethyl thiophenol, o-ethylbenzenethiol, fema no. 3345, ethylbenzenethiol, o, 2-ethylphenylmercaptan, benzenethiol, o-ethyl, o-ethylthiophenol
CCC1=CC=CC=C1S
138.228
138.23
≥95.0% (GC)

Chemical Identifiers

4500-58-7
138.228
ABROBCBIIWHVNS-UHFFFAOYSA-N
3734338
CCC1=CC=CC=C1S
C8H10S
MFCD00010022
2-ethylthiophenol, benzenethiol, 2-ethyl, unii-62gbu45i5p, 2-ethyl thiophenol, o-ethylbenzenethiol, fema no. 3345, ethylbenzenethiol, o, 2-ethylphenylmercaptan, benzenethiol, o-ethyl, o-ethylthiophenol
2-ethylbenzenethiol
Safety and Handling

Safety and Handling

TSCA : Yes

Documents