Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethylbenzeneboronic acid, 98+%

$39.88 - $109.28

Chemical Identifiers

Chemical Name or Material 2-Ethylbenzeneboronic acid
CAS 90002-36-1
Molecular Formula C8H11BO2
Formula Weight 149.99
MDL Number MFCD02093075
Beilstein 2963801
Synonym 2-Ethylphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1771903

 
alfa aesar™
L1771903
1g Each for $39.88

AAL1771906

 
alfa aesar™
L1771906
5g Each for $109.28
Description & Specifications

Specifications

Formula Weight 149.99
Assay ≥98%
Molecular Formula C8H11BO2
MDL Number MFCD02093075
CAS 90002-36-1
Beilstein 2963801
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Chemical Name or Material 2-Ethylbenzeneboronic acid
Synonym 2-Ethylphenylboronic acid
Melting Point 96° to 98°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.