missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylanisole, TCI America™

Manufacturer:  TCI America E050925G

 View more versions of this product

Catalog No. E050925G



Specifications

Specifications

2-Ethylanisole
Yellow
14804-32-1
MFCD00142909
NIEHEMAZEULEKB-UHFFFAOYSA-N
1-ethyl-2-methoxybenzene
84654
Clear Liquid at 20°C
187°C
25g
C9H12O
2-Ethylmethoxybenzene
CCC1=CC=CC=C1OC
136.194
136.19
≥98.0% (GC)

Chemical Identifiers

14804-32-1
136.194
NIEHEMAZEULEKB-UHFFFAOYSA-N
84654
CCC1=CC=CC=C1OC
C9H12O
MFCD00142909
2-Ethylmethoxybenzene
1-ethyl-2-methoxybenzene
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0578&(3)-1035

TSCA : No

Documents