missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylaniline 98%, ACROS Organics™

Chemical Identifiers

CAS 578-54-1
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H11N
Linear Formula C2H5C6H4NH2
Formula Weight 121.18
MDL Number MFCD00007751
Chemical Name or Material 2-Ethylaniline, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC118050010 SDS Thermo Fisher Scientific
118050010
Glass bottle 1kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC118050050 SDS Thermo Fisher Scientific
118050050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC118052500 SDS Thermo Fisher Scientific
118052500
Glass bottle 250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

578-54-1
100.0
C8H11N
121.18
2-Ethylaniline, 98%
97.5
98%
C2H5C6H4NH2
MFCD00007751
≥97.5%

Specifications

578-54-1
100.0
Authentic
1.5590 to 1.5610 (20°C, 589nm)
98%
C8H11N
C2H5C6H4NH2
MFCD00007751
2-Ethylaniline, 98%
97.5
0.98g/mL
Glass bottle
Liquid
Clear yellow to red-brownish
1kg
121.18
12, II, 584
≥97.5%

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-424-2

RTECSNumber : BX9800000

TSCA : TSCA