Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethylamino-3-methoxypyridine, 99%

$79.24 - $79.24

Chemical Identifiers

Chemical Name or Material 2-Ethylamino-3-methoxypyridine
CAS 912761-77-4
Molecular Formula C8H12N2O
Formula Weight 152.2
MDL Number MFCD08061118
Synonym N-Ethyl-3-methoxy-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51013MD

  SDS
alfa aesar™
H51013MD
250mg Each for $79.24
Description & Specifications

Specifications

Formula Weight 152.2
Chemical Name or Material 2-Ethylamino-3-methoxypyridine
Assay 99%
Molecular Formula C8H12N2O
MDL Number MFCD08061118
CAS 912761-77-4
Synonym N-Ethyl-3-methoxy-2-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.