missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethyl-6-methyliodobenzene, 98%, ACROS Organics™

Chemical Identifiers

CAS 175277-95-9
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C9H11I
Linear Formula CH3C6H3(I)C2H5
Formula Weight 246.09
MDL Number MFCD00052855
Chemical Name or Material 2-Ethyl-6-methyliodobenzene
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC355040050 SDS Thermo Fisher Scientific
35504-0050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

175277-95-9
100.0
C9H11I
246.09
2-Ethyl-6-methyliodobenzene
97.5
98%
CH3C6H3(I)C2H5
MFCD00052855
≥97.5%

Specifications

246.09
5g
100.0
97.5
1.5900 to 1.5920 (20°C, 589nm)
CH3C6H3(I)C2H5
MFCD00052855
2-Ethyl-6-methyliodobenzene
Liquid
Clear yellow to red-brown
Authentic
Glass bottle
98%
C9H11I
175277-95-9
≥97.5%

Safety and Handling

LIGHT SENSITIVE

TSCA : TSCA