Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethyl-6-methylaniline, 98%

$123.86 - $551.47

Chemical Identifiers

Chemical Name or Material 2-Ethyl-6-methylaniline
CAS 24549-06-2
Molecular Formula C9H13N
Formula Weight 135.21
MDL Number MFCD00007752
Beilstein 2079468
Synonym 2-Amino-3-ethyltoluene; 6-Ethyl-o-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA127960E

  SDS
alfa aesar™
A127960E
2500g Each for $551.47

AAA1279636

  SDS
alfa aesar™
A1279636
500g Each for $123.86
Description & Specifications

Specifications

Formula Weight 135.21
Boiling Point 226° to 228°C
TSCA TSCA
Beilstein 2079468
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point −25°C
CAS 24549-06-2
Synonym 2-Amino-3-ethyltoluene; 6-Ethyl-o-toluidine
Molecular Formula C9H13N
Quantity 2500g
RTECS Number BY5600000
Assay 98%
MDL Number MFCD00007752
Health Hazard 2 GHS H Statement
H301-H311-H227-H332
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Chemical Name or Material 2-Ethyl-6-methylaniline
Flash Point 89°C (192°F)
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H227-H332
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.