missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Ethyl-5-nitroaniline 98.0+%, TCI America™
$179.17 - $637.06
Chemical Identifiers
CAS | 20191-74-6 |
---|---|
Molecular Formula | C8H10N2O2 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD00034056 |
InChI Key | MMZWMCKTKJKIMC-UHFFFAOYSA-N |
PubChem CID | 4155478 |
IUPAC Name | 2-ethyl-5-nitroaniline |
SMILES | CCC1=C(C=C(C=C1)[N+](=O)[O-])N |
Chemical Identifiers
20191-74-6 | |
166.18 | |
MMZWMCKTKJKIMC-UHFFFAOYSA-N | |
2-ethyl-5-nitroaniline |
C8H10N2O2 | |
MFCD00034056 | |
4155478 | |
CCC1=C(C=C(C=C1)[N+](=O)[O-])N |
Specifications
20191-74-6 | |
C8H10N2O2 | |
1 g | |
CCC1=C(C=C(C=C1)[N+](=O)[O-])N | |
166.18 | |
166.18 | |
Crystalline Powder |
60°C | |
MFCD00034056 | |
MMZWMCKTKJKIMC-UHFFFAOYSA-N | |
2-ethyl-5-nitroaniline | |
4155478 | |
≥98.0% (GC) | |
2-Ethyl-5-nitroaniline |
Safety and Handling
TSCA : No