missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethyl-4-methylthiazole, 98%

Manufacturer:  Alfa Aesar™ A1564214

 View more versions of this product

Catalog No. AAA1564214



Specifications

Specifications

2-Ethyl-4-methylthiazole
160°C to 162°C
15679-12-6
MFCD00036621
2-ethyl-4-methylthiazole, 2-ethyl-4-methyl thiazole, thiazole, 2-ethyl-4-methyl, unii-lvc1zv6x4u, 4-methyl-2-ethylthiazole, lvc1zv6x4u, 2-ethyl-4-methyl-thiazole, fema no. 3680, pubchem9939, acmc-1buw7
CCC1=NC(=CS1)C
127.205
127.21
54°C (130°F)
25g
C6H9NS
UN1993
VGRVKVGGUPOCMT-UHFFFAOYSA-N
2-ethyl-4-methyl-1,3-thiazole
27440
98%

Chemical Identifiers

15679-12-6
127.205
VGRVKVGGUPOCMT-UHFFFAOYSA-N
27440
CCC1=NC(=CS1)C
C6H9NS
MFCD00036621
2-ethyl-4-methylthiazole, 2-ethyl-4-methyl thiazole, thiazole, 2-ethyl-4-methyl, unii-lvc1zv6x4u, 4-methyl-2-ethylthiazole, lvc1zv6x4u, 2-ethyl-4-methyl-thiazole, fema no. 3680, pubchem9939, acmc-1buw7
2-ethyl-4-methyl-1,3-thiazole
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

Documents