Please login to your online account to display your discounted pricing

2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde
CAS 88634-80-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H10N2O
Formula Weight 138.17
MDL Number MFCD00173727
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01365EA

  SDS
fisher scientific company
SB01365EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 138.17
Quantity 10g
Assay 97%
CAS Min % 97.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Assay Percent Range 97%
MDL Number MFCD00173727
Chemical Name or Material 2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde
Packaging Amber glass bottle
CAS 88634-80-4
Molecular Formula C7H10N2O
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.