missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethyl-4,5-dimethylthiazole, TCI America™

Manufacturer:  TCI America E07185G

 View more versions of this product

Catalog No. E07185G



Specifications

Specifications

2-Ethyl-4,5-dimethylthiazole
Yellow
873-64-3
MFCD00053125
CCC1=NC(=C(S1)C)C
141.232
141.23
≥98.0% (GC)
185°C
5g
C7H11NS
BVOKJOZBJOWZNJ-UHFFFAOYSA-N
2-ethyl-4,5-dimethyl-1,3-thiazole
522877
Clear Liquid at 20°C

Chemical Identifiers

873-64-3
141.232
BVOKJOZBJOWZNJ-UHFFFAOYSA-N
2-ethyl-4,5-dimethyl-1,3-thiazole
C7H11NS
MFCD00053125
522877
CCC1=NC(=C(S1)C)C
Safety and Handling

Safety and Handling

TSCA : No

Documents