Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethyl-4,5-dimethyloxazole, 98%

$35.41 - $98.82

Chemical Identifiers

Chemical Name or Material 2-Ethyl-4,5-dimethyloxazole
CAS 53833-30-0
Molecular Formula C7H11NO
Formula Weight 125.18
MDL Number MFCD00036664
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2747003

 
alfa aesar™
H2747003
1g Each for $35.41

AAH2747006

 
alfa aesar™
H2747006
5g Each for $98.82
Description & Specifications

Specifications

Formula Weight 125.18
Chemical Name or Material 2-Ethyl-4,5-dimethyloxazole
TSCA TSCA
Assay 98%
Molecular Formula C7H11NO
MDL Number MFCD00036664
CAS 53833-30-0
Quantity 1g