Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethyl-4,5-dimethyl-3-thiazoline, cis + trans, 99%

$44.95 - $226.27

Chemical Identifiers

Chemical Name or Material 2-Ethyl-4,5-dimethyl-3-thiazoline
CAS 76788-46-0
Name Note Cis + Trans
Molecular Formula C7H13NS
Formula Weight 143.26
MDL Number MFCD01680822
Synonym 4,5-Dimethyl-2-ethyl-3-thiazoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2696003

 
alfa aesar™
H2696003
1g Each for $44.95

AAH2696009

 
alfa aesar™
H2696009
10g Each for $226.27
Description & Specifications

Specifications

Formula Weight 143.26
Chemical Name or Material 2-Ethyl-4,5-dimethyl-3-thiazoline
Name Note Cis + Trans
Assay 99%
Molecular Formula C7H13NS
MDL Number MFCD01680822
CAS 76788-46-0
Synonym 4,5-Dimethyl-2-ethyl-3-thiazoline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 65°C (3mmHg)
Quantity 1g
Flash Point 65°C (149°F)
Recommended Storage 2° to 8°C
RTECS Number XJ6642500
Safety and Handling

Warning

GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.

2° to 8°C