Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethyl-3-methoxypyrazine, 99%

$64.80 - $163.16

Chemical Identifiers

Chemical Name or Material 2-Ethyl-3-methoxypyrazine
CAS 25680-58-4
Molecular Formula C7H10N2O
Formula Weight 138.17
MDL Number MFCD00006129
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1581706

  SDS
alfa aesar™
A1581706
5g Each for $163.16

AAA1581703

  SDS
alfa aesar™
A1581703
1g Each for $64.80
Description & Specifications

Specifications

Formula Weight 138.17
Chemical Name or Material 2-Ethyl-3-methoxypyrazine
TSCA TSCA
Assay 99%
Molecular Formula C7H10N2O
MDL Number MFCD00006129
CAS 25680-58-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 90°C (40mmHg)
Quantity 5g
Flash Point 69°C (156°F)
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.