Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethyl-3-methoxybenzoic acid, 98%

Chemical Identifiers

Chemical Name or Material 2-Ethyl-3-methoxybenzoic acid
CAS 57598-51-3
Formula Weight 180.2
MDL Number MFCD11100772
Beilstein 2804017
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6645403

 
alfa aesar™
H6645403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6645406

 
alfa aesar™
H6645406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66454MD

 
alfa aesar™
H66454MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.2
Physical Form Solid
Assay 98%
MDL Number MFCD11100772
CAS 57598-51-3
Chemical Name or Material 2-Ethyl-3-methoxybenzoic acid
Beilstein 2804017
Storage Note 1 -30° to -10°C
Quantity 1g
Safety and Handling

-30° to -10°C