Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethyl-2-vinyl-1,3-dioxolane, 98%

Chemical Identifiers

Chemical Name or Material 2-Ethyl-2-vinyl-1,3-dioxolane
CAS 22515-82-8
Molecular Formula C7H12O2
Formula Weight 128.17
MDL Number MFCD13185932
UN Number UN3271
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3197109

  SDS
alfa aesar™
H3197109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3197118

  SDS
alfa aesar™
H3197118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.17
Chemical Name or Material 2-Ethyl-2-vinyl-1,3-dioxolane
Assay 98%
Molecular Formula C7H12O2
MDL Number MFCD13185932
CAS 22515-82-8
UN Number UN3271
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 70°C (77mmHg)
Odor Characteristic
Quantity 10g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

Applications
It is an important raw material in organic synthesis, agricultural fields and in dyestuffs.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.