missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethyl-2-oxazoline, TCI America™

Manufacturer:  TCI America E0373100G

 View more versions of this product

Catalog No. E0373100G



Specifications

Specifications

2-Ethyl-2-oxazoline
Colorless
10431-98-8
MFCD00005302
2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
CCC1=NCCO1
99.133
99.13
≥98.0% (GC,T)
128°C
100g
C5H9NO
1993
NYEZZYQZRQDLEH-UHFFFAOYSA-N
2-ethyl-4,5-dihydro-1,3-oxazole
66412
Clear Liquid at 20°C

Chemical Identifiers

10431-98-8
99.133
NYEZZYQZRQDLEH-UHFFFAOYSA-N
66412
CCC1=NCCO1
C5H9NO
MFCD00005302
2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
2-ethyl-4,5-dihydro-1,3-oxazole
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5627

TSCA : Yes

Documents