missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethyl-2-oxazoline, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 159985000

 View more versions of this product

Catalog No. AC159985000



Specifications

Specifications

2-Ethyl-2-oxazoline
29°C
Glass bottle
0.98
0.2% max.
Undesignated
500g
99% min. (GC)
2-ethyl-2-oxazoline
NYEZZYQZRQDLEH-UHFFFAOYSA-N
2-ethyl-4,5-dihydro-1,3-oxazole
66412
Liquid
0.9800g/mL
Authentic
1.4360 to 1.4380
0.88 mPa.s (20°C)
128.4°C
-62.0°C
10431-98-8
C5H9NO
Solubility in water: miscible
CCC1=NCCO1
99.13
99.13
99+%

Chemical Identifiers

10431-98-8
99.13
2-ethyl-2-oxazoline
2-ethyl-4,5-dihydro-1,3-oxazole
C5H9NO
NYEZZYQZRQDLEH-UHFFFAOYSA-N
66412
CCC1=NCCO1
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta

GHS Signal Word: Danger

EINECSNumber : 233-912-4

SDS Documents