Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethyl-2-oxazoline, 99%

Chemical Identifiers

Chemical Name or Material 2-Ethyl-2-oxazoline
CAS 10431-98-8
Molecular Formula C5H9NO
Formula Weight 99.13
MDL Number MFCD00005302
UN Number UN2734
Beilstein 105464
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2328722

 
alfa aesar™
B2328722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 99.13
Chemical Name or Material 2-Ethyl-2-oxazoline
TSCA TSCA
Assay 99%
Molecular Formula C5H9NO
MDL Number MFCD00005302
CAS 10431-98-8
UN Number UN2734
Beilstein 105464
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
Flash Point 29°C (84°F)
Odor Amine-like
Boiling Point 128° to 129°C
Melting Point −62°C
Quantity 100g
pH 10 to 12

Applications
2-ethyl-2-oxazoline is an organic intermediate and monomer that is used in water-soluble polymers, pharmaceuticals, cosmetics, specialty coatings and adhesives.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store away from air, oxidizing agents. Store under dry inert gas.

Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H226-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
May cause an allergic skin reaction.