Please login to your online account to display your discounted pricing

2-Ethyl-2-methyl-1,3-dioxolane, 99%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  271971000

 View more versions of this product

Catalog No. AC271971000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 116.16
Quantity 100mL
Color Clear colorless
EINECS Number 204-785-2
TSCA TSCA
Density 0.9300g/mL
RTECS Number JH7700000
CAS Max % 100.0
Chemical Name or Material 2-Ethyl-2-methyl-1, 3-dioxolane
Packaging Glass bottle
Refractive Index 1.4080 to 1.4100 (20°C, 589nm)
Name Note 99%
Molecular Formula C6H12O2
Synonym 2-Butanone ethylene acetal
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 99%
MDL Number MFCD00014108
CAS 126-39-6

Specifications

Chemical Name or Material 2-Ethyl-2-methyl-1, 3-dioxolane
Name Note 99%
CAS 126-39-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H12O2
Formula Weight 116.16
MDL Number MFCD00014108
Synonym 2-Butanone ethylene acetal
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents