missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 164655000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Ethyl-2-(hydroxymethyl)-1, 3-propanediol | |
0.002% max. (as formic acid) | |
100.0 | |
172°C | |
98% | |
C6H14O3 | |
MFCD00004694 | |
01, III, 2349 | |
Solubility in water: soluble. Other solubilities: freely soluble in alcohols,insoluble in hydrocarbons | |
CCC(CO)(CO)CO | |
134.18 | |
157 mPa.s (75°C) | |
98% |
77-99-6 | |
97.5 | |
160°C (0.7 hPa) | |
Authentic | |
Plastic bottle | |
C2H5C(CH2OH)3 | |
500g | |
trimethylolpropane, 2-ethyl-2-hydroxymethyl propane-1,3-diol, ethriol, 2-ethyl-2-hydroxymethyl-1,3-propanediol, hexaglycerine, ettriol, etriol, trimethylol propane, tmp alcohol, 1,1,1-tris hydroxymethyl propane | |
ZJCCRDAZUWHFQH-UHFFFAOYSA-N | |
2-ethyl-2-(hydroxymethyl)propane-1,3-diol | |
6510 | |
134.18 |
Chemical Identifiers
77-99-6 | |
134.18 | |
ZJCCRDAZUWHFQH-UHFFFAOYSA-N | |
6510 | |
CCC(CO)(CO)CO |
C6H14O3 | |
MFCD00004694 | |
trimethylolpropane, 2-ethyl-2-hydroxymethyl propane-1,3-diol, ethriol, 2-ethyl-2-hydroxymethyl-1,3-propanediol, hexaglycerine, ettriol, etriol, trimethylol propane, tmp alcohol, 1,1,1-tris hydroxymethyl propane | |
2-ethyl-2-(hydroxymethyl)propane-1,3-diol |
Safety and Handling
EINECSNumber : 201-074-9
RUO â Research Use Only