missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethyl-2-hexenal (cis- and trans- mixture), TCI America™

Manufacturer:  TCI America E0123100ML

 View more versions of this product

Catalog No. E0123100ML



Specifications

Specifications

2-Ethyl-2-hexenal (cis- and trans- mixture)
Yellow
645-62-5
MFCD00006983
CCCC=C(CC)C=O
126.199
126.20
≥94.0% (GC)
175°C
100mL
C8H14O
PYLMCYQHBRSDND-VURMDHGXSA-N
(Z)-2-ethylhex-2-enal
10997035
Clear Liquid at 20°C

Chemical Identifiers

645-62-5
126.199
PYLMCYQHBRSDND-VURMDHGXSA-N
(Z)-2-ethylhex-2-enal
C8H14O
MFCD00006983
10997035
CCCC=C(CC)C=O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0513

RTECSNumber : MP6300000

TSCA : Yes

Documents