Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethyl-1H-indene, 97%

$306.31 - $1,172.93

Chemical Identifiers

Chemical Name or Material 2-Ethyl-1H-indene
CAS 17059-50-6
Molecular Formula C11H12
Formula Weight 144.22
MDL Number MFCD01321239
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2310406

 
alfa aesar™
B2310406
5g Each for $306.31

AAB2310414

 
alfa aesar™
B2310414
25g Each for $1,172.93
Description & Specifications

Specifications

Formula Weight 144.22
Chemical Name or Material 2-Ethyl-1H-indene
Assay 97%
Molecular Formula C11H12
MDL Number MFCD01321239
CAS 17059-50-6
Boiling Point 94° to 96°C (10mmHg)
Health Hazard 1 Warning
Quantity 5g
Flash Point 87°C (190°F)
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.