Please login to your online account to display your discounted pricing

2-Ethyl-1-hexanol, 99%, ACROS Organics™


Quantity 5mL
Packaging Glass Bottle

Manufacturer: fisher scientific acros  118530050

 View more versions of this product

Catalog No. AC118530050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 130.23
Quantity 5mL
Color Clear
Merck Index 15, 3863
Packaging Glass Bottle
Density 0.8330g/mL
CAS Max % 100.0
Molecular Formula C8H18O
Linear Formula CH3(CH2)3CH(C2H5)CH2OH
MDL Number MFCD00004746
CAS 104-76-7
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Ethyl-1-hexanol
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4300 to 1.4330 (20°C, 589nm)
Water 0.1% max.
Health Hazard 1 Warning

Specifications

Chemical Name or Material 2-Ethyl-1-hexanol
CAS 104-76-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H18O
Linear Formula CH3(CH2)3CH(C2H5)CH2OH
Formula Weight 130.23
MDL Number MFCD00004746
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents