missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethyl-1-hexanethiol, TCI America™

Manufacturer:  TCI America E1031100G

 View more versions of this product

Catalog No. E1031100G



Specifications

Specifications

2-Ethyl-1-hexanethiol
Colorless
100g
C8H18S
3336
UCJMHYXRQZYNNL-UHFFFAOYSA-N
2-ethylhexane-1-thiol
110968
Clear Liquid at 20°C
67°C
-40°C
7341-17-5
MFCD00039654
2-Ethylhexyl Mercaptan
CCCCC(CC)CS
146.292
146.29
≥98.0% (GC)

Chemical Identifiers

7341-17-5
146.292
UCJMHYXRQZYNNL-UHFFFAOYSA-N
110968
CCCCC(CC)CS
C8H18S
MFCD00039654
2-Ethylhexyl Mercaptan
2-ethylhexane-1-thiol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0464

TSCA : Yes

Documents