Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethyl-1-butylamine, 97%

Chemical Identifiers

Chemical Name or Material 2-Ethyl-1-butylamine
CAS 617-79-8
Molecular Formula C6H15N
Linear Formula (CH3CH2)2CHCH2NH2
Formula Weight 101.19g/mol
MDL Number MFCD00040755
UN Number UN3286
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2085509

 
alfa aesar™
B20855-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2085588

 
alfa aesar™
B20855-88
2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 125°C
Quantity 10g
Flash Point 14°C (57°F)
Odor Amine-like
Chemical Name or Material 2-Ethyl-1-butylamine
CAS 617-79-8
Assay 97%
Molecular Formula C6H15N
Linear Formula (CH3CH2)2CHCH2NH2
Formula Weight 101.19g/mol
MDL Number MFCD00040755
UN Number UN3286
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H331-H314-H318-H302-H312
Highly flammable liquid and vapour.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
RTECS Number EO4900000
Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H225-H331-H314-H318-H302-H312
Highly flammable liquid and vapour.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.