Please login to your online account to display your discounted pricing

2-Ethyl-1-butanol 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  118170050

 View more versions of this product

Catalog No. AC118170050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 102.18
Quantity 5g
TSCA TSCA
RTECS Number EL3850000
EINECS Number 202-621-4
Density 0.8300g/mL
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Synonym 2-Ethylbutyl alcohol
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Linear Formula (C2H5)2CHCH2OH
Molecular Formula C6H14O
Chemical Name or Material 2-Ethyl-1-butanol
Assay Percent Range 98%
CAS 97-95-0
Name Note 98%
MDL Number MFCD00004744

Specifications

Chemical Name or Material 2-Ethyl-1-butanol
Name Note 98%
CAS 97-95-0
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C6H14O
Linear Formula (C2H5)2CHCH2OH
Formula Weight 102.18
MDL Number MFCD00004744
Synonym 2-Ethylbutyl alcohol
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents