missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethyl-1-butanol 98%, ACROS Organics™

Chemical Identifiers

CAS 97-95-0
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C6H14O
Linear Formula (C2H5)2CHCH2OH
Formula Weight 102.18
MDL Number MFCD00004744
Synonym 2-Ethylbutyl alcohol
Chemical Name or Material 2-Ethyl-1-butanol, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC118170050 SDS Thermo Fisher Scientific
118170050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97-95-0
100.0
C6H14O
102.18
2-Ethylbutyl alcohol
97.5
98%
(C2H5)2CHCH2OH
MFCD00004744
2-Ethyl-1-butanol, 98%

Specifications

102.18
0.8300g/mL
97.5
Glass bottle
C6H14O
97-95-0
2-Ethyl-1-butanol, 98%
5g
100.0
2-Ethylbutyl alcohol
(C2H5)2CHCH2OH
98%
MFCD00004744

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-621-4

RTECSNumber : EL3850000

TSCA : TSCA