Please login to your online account to display your discounted pricing

2-Ethyl-1,3-dithiane +97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Ethyl-1, 3-dithiane
CAS 6007-23-4
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C6H12S2
Formula Weight 148.29
MDL Number MFCD12910519
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC393100050

  SDS
thermo fisher scientific
AC393100050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.0%
Formula Weight 148.29
Physical Form Liquid
Quantity 5g
Color Clear colorless to slightly yellow
CAS Max % 100.0
CAS Min % 97.0
Beilstein 19, V,1, 70
Packaging Glass bottle
Molecular Formula C6H12S2
CAS 6007-23-4
MDL Number MFCD12910519
Refractive Index 1.5450 to 1.5470 (20°C, 589nm)
Infrared Spectrum Authentic
Chemical Name or Material 2-Ethyl-1, 3-dithiane
Assay Percent Range 97+%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.