missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethoxythiazole, TCI America™

Manufacturer:  TCI America E066925G

 View more versions of this product

Catalog No. E066925G



Specifications

Specifications

2-Ethoxythiazole
Yellow
15679-19-3
NDUWJHRKDYXRAD-UHFFFAOYSA-N
2-ethoxy-1,3-thiazole
61809
Clear Liquid at 20°C
160°C
25g
C5H7NOS
CCOC1=NC=CS1
129.177
129.18
≥98.0% (GC)

Chemical Identifiers

15679-19-3
129.177
61809
CCOC1=NC=CS1
C5H7NOS
NDUWJHRKDYXRAD-UHFFFAOYSA-N
2-ethoxy-1,3-thiazole
Safety and Handling

Safety and Handling

TSCA : No

Documents