missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Ethoxypyridine 98.0+%, TCI America™
$28.21 - $91.24
Chemical Identifiers
CAS | 14529-53-4 |
---|---|
Molecular Formula | C7H9NO |
Molecular Weight (g/mol) | 123.16 |
MDL Number | MFCD00023464 |
InChI Key | LISKAOIANGDBTB-UHFFFAOYSA-N |
Synonym | pyridine, 2-ethoxy, 2-ethoxy pyridine, 2-ethoxy-pyridine, zlchem 304, pubchem1126, acmc-209cun, ethyl 2-pyridinyl ether, ksc497c7d |
PubChem CID | 84499 |
IUPAC Name | 2-ethoxypyridine |
SMILES | CCOC1=CC=CC=N1 |
Chemical Identifiers
14529-53-4 | |
123.16 | |
LISKAOIANGDBTB-UHFFFAOYSA-N | |
84499 | |
CCOC1=CC=CC=N1 |
C7H9NO | |
MFCD00023464 | |
pyridine, 2-ethoxy, 2-ethoxy pyridine, 2-ethoxy-pyridine, zlchem 304, pubchem1126, acmc-209cun, ethyl 2-pyridinyl ether, ksc497c7d | |
2-ethoxypyridine |
Specifications
14529-53-4 | |
157°C | |
MFCD00023464 | |
1993 | |
LISKAOIANGDBTB-UHFFFAOYSA-N | |
2-ethoxypyridine | |
84499 | |
≥98.0% (GC,T) | |
2-Ethoxypyridine |
Yellow | |
C7H9NO | |
5 g | |
pyridine, 2-ethoxy, 2-ethoxy pyridine, 2-ethoxy-pyridine, zlchem 304, pubchem1126, acmc-209cun, ethyl 2-pyridinyl ether, ksc497c7d | |
CCOC1=CC=CC=N1 | |
123.16 | |
123.16 | |
Liquid |
Safety and Handling
TSCA : No