Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxypyridine-3-carboxaldehyde, 97%

$232.43 - $232.43

Chemical Identifiers

Chemical Name or Material 2-Ethoxypyridine-3-carboxaldehyde
CAS 885278-07-9
Molecular Formula C8H9NO2
Formula Weight 151.17
MDL Number MFCD06804559
Synonym 2-Ethoxynicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5008803

 
alfa aesar™
H5008803
1g Each for $232.43
Description & Specifications

Specifications

Formula Weight 151.17
Chemical Name or Material 2-Ethoxypyridine-3-carboxaldehyde
Assay 97%
Molecular Formula C8H9NO2
MDL Number MFCD06804559
CAS 885278-07-9
Synonym 2-Ethoxynicotinaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
Used as pharmaceutical intermediate.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.