Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxypyrazine, 98%

$74.51 - $618.80

Chemical Identifiers

Chemical Name or Material 2-Ethoxypyrazine
CAS 38028-67-0
Molecular Formula C6H9N2O
Formula Weight 125.15
MDL Number MFCD00014609
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2521906

 
alfa aesar™
B2521906
5g Each for $74.51

AAB2521914

 
alfa aesar™
B2521914
25g Each for $209.34

AAB2521922

 
alfa aesar™
B2521922
100g Each for $618.80
Description & Specifications

Specifications

Formula Weight 125.15
Chemical Name or Material 2-Ethoxypyrazine
Assay 98%
Molecular Formula C6H9N2O
MDL Number MFCD00014609
CAS 38028-67-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.