Please login to your online account to display your discounted pricing

2-Ethoxyphenylboronic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Ethoxyphenylboronic acid
CAS 213211-69-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H11BO3
Linear Formula C2H5OC6H4B(OH)2
Formula Weight 165.98
MDL Number MFCD00674027
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC377670050

  SDS
thermo fisher scientific
AC377670050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC377670010

  SDS
thermo fisher scientific
AC377670010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC377670100

  SDS
thermo fisher scientific
AC377670100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder or Crystals
Assay ≥97.5%
Formula Weight 165.98
Quantity 5g
Color White to off-white
Melting Point 97° to 102°C
CAS Max % 100.0
Chemical Name or Material 2-Ethoxyphenylboronic acid
Packaging Glass bottle
CAS 213211-69-9
Assay Percent Range 98%
Molecular Formula C8H11BO3
Linear Formula C2H5OC6H4B(OH)2
MDL Number MFCD00674027
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.