missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethoxyphenol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC370031000

 View more versions of this product

Catalog No. AC370031000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Liquid
Clear colorless to yellow
100.0
2-Ethoxyphenol
97.5
Glass bottle
C2H5OC6H4OH
98%
138.17
100g
1.0900g/mL
Authentic
MFCD00002187
1.5280 to 1.5300 (20°C, 589nm)
Pyrocatechol monoethyl ether
C8H10O2
94-71-3

Chemical Identifiers

94-71-3
100.0
C8H10O2
138.17
Pyrocatechol monoethyl ether
≥97.5%
97.5
98%
C2H5OC6H4OH
MFCD00002187
2-Ethoxyphenol

Safety and Handling

SDS & Safety Documents

GHS H Statement
May cause an allergic skin reaction. Causes serious eye damage. Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Gently wash with plenty of soap and water. Avoid release to the environment.

Danger