Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxynicotinic acid, 98%

$67.81 - $842.28

Chemical Identifiers

Chemical Name or Material 2-Ethoxynicotinic acid
CAS 35969-54-1
Molecular Formula C8H9NO3
Formula Weight 167.16
MDL Number MFCD00128788
Synonym 2-Ethoxypyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2088709

  SDS
alfa aesar™
B2088709
10g Each for $67.81

AAB2088718

  SDS
alfa aesar™
B2088718
50g Each for $192.86

AAB2088730

  SDS
alfa aesar™
B2088730
250g Each for $842.28
Description & Specifications

Specifications

Formula Weight 167.16
Quantity 10g
MDL Number MFCD00128788
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C8H9NO3
CAS 35969-54-1
Chemical Name or Material 2-Ethoxynicotinic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 2-Ethoxypyridine-3-carboxylic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.