missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethoxynaphthoic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 243000100

 View more versions of this product

Catalog No. AC243000100



Specifications

Specifications

2-Ethoxynaphthoic acid
Glass bottle
141°C to 145°C
98%
97.5
98%
MFCD00004008
MYFBSSDLYGWAHH-UHFFFAOYSA-N
2-ethoxynaphthalene-1-carboxylic acid
75210
Crystalline Powder
Authentic
Brown to White
10g
2224-00-2
100.0
C13H12O3
1-naphthalenecarboxylic acid, 2-ethoxy, 2-ethoxy-1-naphthoic acid, 2-ethoxy-1-naphthoicacid, 2-ethoxynaphthalenecarboxylic acid, 2-ethoxynaphthoic acid, 2-ethoxynaphtoic acid, acmc-209ftv, rarechem al be 0566, timtec-bb sbb005771
CCOC1=C(C2=CC=CC=C2C=C1)C(=O)O
216.24
216.24
98%

Chemical Identifiers

2224-00-2
216.24
MYFBSSDLYGWAHH-UHFFFAOYSA-N
75210
CCOC1=C(C2=CC=CC=C2C=C1)C(=O)O
C13H12O3
MFCD00004008
1-naphthalenecarboxylic acid, 2-ethoxy, 2-ethoxy-1-naphthoic acid, 2-ethoxy-1-naphthoicacid, 2-ethoxynaphthalenecarboxylic acid, 2-ethoxynaphthoic acid, 2-ethoxynaphtoic acid, acmc-209ftv, rarechem al be 0566, timtec-bb sbb005771
2-ethoxynaphthalene-1-carboxylic acid
Safety and Handling

Safety and Handling

EINECSNumber : 218-745-7

TSCA : TSCA

SDS
Documents