Please login to your online account to display your discounted pricing

2-Ethoxynaphthoic acid, 98%, ACROS Organics™

$32.07 - $32.07

Chemical Identifiers

Chemical Name or Material 2-Ethoxynaphthoic acid
Name Note 98%
CAS 2224-00-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H12O3
Formula Weight 216.24
MDL Number MFCD00004008
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC243000100

  SDS
thermo fisher scientific
243000100
Glass bottle 10g Each for $32.07
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 216.24
Quantity 10g
Color White to pale brown
Melting Point 141° to 145°C
EINECS Number 218-745-7
TSCA TSCA
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 97.5
Packaging Glass bottle
Synonym 2-Ethoxynaphthalene-1-carboxylic acid
CAS 2224-00-2
Molecular Formula C13H12O3
Chemical Name or Material 2-Ethoxynaphthoic acid
Name Note 98%
MDL Number MFCD00004008
Assay Percent Range 98%