missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethoxynaphthalene, TCI America™

$19.86 - $70.28

Chemical Identifiers

CAS 93-18-5
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.227
MDL Number MFCD00016808
InChI Key GUMOJENFFHZAFP-UHFFFAOYSA-N
Synonym bromelia, naphthalene, 2-ethoxy, neroline, nerolin new, nerolin ii, ethyl 2-naphthyl ether, 2-naphthol ethyl ether, bromelia compound, beta-naphthyl ethyl ether, ethyl beta-naphthyl ether
PubChem CID 7129
IUPAC Name 2-ethoxynaphthalene
SMILES CCOC1=CC2=CC=CC=C2C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0431100G View Documents TCI America
N0431100G
100g Each for $70.28
 
N043125G View Documents TCI America
N043125G
25g Each for $19.86
 
Specifications

Chemical Identifiers

93-18-5
172.227
GUMOJENFFHZAFP-UHFFFAOYSA-N
7129
CCOC1=CC2=CC=CC=C2C=C1
C12H12O
MFCD00016808
bromelia, naphthalene, 2-ethoxy, neroline, nerolin new, nerolin ii, ethyl 2-naphthyl ether, 2-naphthol ethyl ether, bromelia compound, beta-naphthyl ethyl ether, ethyl beta-naphthyl ether
2-ethoxynaphthalene

Specifications

282°C
35°C
93-18-5
MFCD00016808
GUMOJENFFHZAFP-UHFFFAOYSA-N
2-ethoxynaphthalene
7129
Crystal-Powder at 20°C
2-Ethoxynaphthalene
White
100g
C12H12O
bromelia, naphthalene, 2-ethoxy, neroline, nerolin new, nerolin ii, ethyl 2-naphthyl ether, 2-naphthol ethyl ether, bromelia compound, beta-naphthyl ethyl ether, ethyl beta-naphthyl ether
CCOC1=CC2=CC=CC=C2C=C1
172.227
172.23
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0362

RTECSNumber : QJ6900000

TSCA : Yes