Learn More
2-Ethoxynaphthalene, 99%, Thermo Scientific Chemicals
$35.01 - $182.42
Chemical Identifiers
CAS | 93-18-5 |
---|---|
Molecular Formula | C12H12O |
Molecular Weight (g/mol) | 172.227 |
MDL Number | MFCD00016808 |
InChI Key | GUMOJENFFHZAFP-UHFFFAOYSA-N |
Synonym | bromelia, naphthalene, 2-ethoxy, neroline, nerolin new, nerolin ii, ethyl 2-naphthyl ether, 2-naphthol ethyl ether, bromelia compound, beta-naphthyl ethyl ether, ethyl beta-naphthyl ether |
PubChem CID | 7129 |
IUPAC Name | 2-ethoxynaphthalene |
SMILES | CCOC1=CC2=CC=CC=C2C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2332714
|
Thermo Scientific Chemicals
B2332714 |
25 g |
Each for $35.01
|
|
|||||
AAB2332722
|
Thermo Scientific Chemicals
B2332722 |
100 g |
Each for $70.71
|
|
|||||
AAB2332736
|
Thermo Scientific Chemicals
B2332736 |
500 g |
Each for $182.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93-18-5 | |
172.227 | |
GUMOJENFFHZAFP-UHFFFAOYSA-N | |
7129 | |
CCOC1=CC2=CC=CC=C2C=C1 |
C12H12O | |
MFCD00016808 | |
bromelia, naphthalene, 2-ethoxy, neroline, nerolin new, nerolin ii, ethyl 2-naphthyl ether, 2-naphthol ethyl ether, bromelia compound, beta-naphthyl ethyl ether, ethyl beta-naphthyl ether | |
2-ethoxynaphthalene |
Specifications
93-18-5 | |
282°C | |
C12H12O | |
25 g | |
14,3752 | |
GUMOJENFFHZAFP-UHFFFAOYSA-N | |
2-ethoxynaphthalene | |
7129 | |
99% |
35°C to 37°C | |
134°C (273°F) | |
MFCD00016808 | |
2042397 | |
bromelia, naphthalene, 2-ethoxy, neroline, nerolin new, nerolin ii, ethyl 2-naphthyl ether, 2-naphthol ethyl ether, bromelia compound, beta-naphthyl ethyl ether, ethyl beta-naphthyl ether | |
CCOC1=CC2=CC=CC=C2C=C1 | |
172.227 | |
172.23 | |
2-Ethoxynaphthalene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 202-226-7
RTECSNumber : QJ6900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only