missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethoxyethyl Methacrylate (stabilized with MEHQ), TCI America™

$38.19 - $117.64

Chemical Identifiers

CAS 2370-63-0
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00009249
InChI Key SFPNZPQIIAJXGL-UHFFFAOYSA-N
Synonym 2-ethoxyethyl methacrylate, ethoxyethyl methacrylate, 2-propenoic acid, 2-methyl-, 2-ethoxyethyl ester, 2-ethoxy ethyl methacrylate, methacrylic acid, 2-ethoxyethyl ester, unii-5q6xr4mr8e, methacrylic acid 2-ethoxyethyl ester, 5q6xr4mr8e, 2-ethoxyethyl 2-methylacrylate, 2-ethyloxy ethyl 2-methylprop-2-enoate
PubChem CID 71404
IUPAC Name 2-ethoxyethyl 2-methylprop-2-enoate
SMILES CCOCCOC(=O)C(=C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M1251500ML View Documents TCI America
M1251500ML
500mL Each for $117.64
 
M125125ML View Documents TCI America
M125125ML
25mL Each for $38.19
 
Specifications

Chemical Identifiers

2370-63-0
158.197
SFPNZPQIIAJXGL-UHFFFAOYSA-N
71404
CCOCCOC(=O)C(=C)C
C8H14O3
MFCD00009249
2-ethoxyethyl methacrylate, ethoxyethyl methacrylate, 2-propenoic acid, 2-methyl-, 2-ethoxyethyl ester, 2-ethoxy ethyl methacrylate, methacrylic acid, 2-ethoxyethyl ester, unii-5q6xr4mr8e, methacrylic acid 2-ethoxyethyl ester, 5q6xr4mr8e, 2-ethoxyethyl 2-methylacrylate, 2-ethyloxy ethyl 2-methylprop-2-enoate
2-ethoxyethyl 2-methylprop-2-enoate

Specifications

93°C
500mL
C8H14O3
2-ethoxyethyl methacrylate, ethoxyethyl methacrylate, 2-propenoic acid, 2-methyl-, 2-ethoxyethyl ester, 2-ethoxy ethyl methacrylate, methacrylic acid, 2-ethoxyethyl ester, unii-5q6xr4mr8e, methacrylic acid 2-ethoxyethyl ester, 5q6xr4mr8e, 2-ethoxyethyl 2-methylacrylate, 2-ethyloxy ethyl 2-methylprop-2-enoate
CCOCCOC(=O)C(=C)C
158.197
158.20
≥98.0% (GC)
Colorless
2370-63-0
MFCD00009249
SFPNZPQIIAJXGL-UHFFFAOYSA-N
2-ethoxyethyl 2-methylprop-2-enoate
71404
Clear Liquid at 20°C
2-Ethoxyethyl Methacrylate (stabilized with MEHQ)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1049

TSCA : Yes