missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethoxyethyl Isobutyrate, TCI America™

$71.81 - $229.17

Chemical Identifiers

CAS 54396-97-3
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.213
MDL Number MFCD00009250
InChI Key ANSOCBLHUNAGRK-UHFFFAOYSA-N
Synonym Cellosolve Isobutyrate, Isobutyric Acid 2-Ethoxyethyl Ester, Ethylene Glycol Monoethyl Ether Isobutyrate
PubChem CID 108587
IUPAC Name 2-ethoxyethyl 2-methylpropanoate
SMILES CCOCCOC(=O)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I02825ML View Documents TCI America
I02825ML
5mL Each for $71.81
 
I028225ML View Documents TCI America
I028225ML
25mL Each for $229.17
 
Specifications

Chemical Identifiers

54396-97-3
160.213
ANSOCBLHUNAGRK-UHFFFAOYSA-N
108587
CCOCCOC(=O)C(C)C
C8H16O3
MFCD00009250
Cellosolve Isobutyrate, Isobutyric Acid 2-Ethoxyethyl Ester, Ethylene Glycol Monoethyl Ether Isobutyrate
2-ethoxyethyl 2-methylpropanoate

Specifications

85°C
5mL
C8H16O3
3272
ANSOCBLHUNAGRK-UHFFFAOYSA-N
2-ethoxyethyl 2-methylpropanoate
108587
Clear Liquid at 20°C
2-Ethoxyethyl Isobutyrate
Yellow
54396-97-3
MFCD00009250
Cellosolve Isobutyrate, Isobutyric Acid 2-Ethoxyethyl Ester, Ethylene Glycol Monoethyl Ether Isobutyrate
CCOCCOC(=O)C(C)C
160.213
160.21
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes