missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethoxyethyl Ether, 98+%, Extra Pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC117900100

 View more versions of this product

Catalog No. AC117900100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Ethoxyethyl ether
82°C
Metal drum
0.05% max.
1.4 mPa.s (20°C)
180°C to 190°C
-44.3°C
112-36-7
100.0
C8H18O3
MFCD00009254
15, 3132
Solubility in water: soluble. Other solubilities: very soluble in alcohol and other organic solvents,soluble in hydrocarbons,soluble in ether
CCOCCOCCOCC
162.23
CHEBI:44664
Liquid
Extra Pure
0.9000g/mL
Authentic
1.4105 to 1.4125
0.9
0.2% max. (K.F.)
Undesignated
10kg
98.0
98% min. (GC)
(C2H5OCH2CH2)2O
01, II, 519
1-ethoxy-2-2-ethoxyethoxy ethane, 2-ethoxyethyl ether, 3,6,9-trioxaundecane, bis 2-ethoxyethyl ether, degdee, diethyl carbitol, diethyldiethylene glycol, diethylene glycol diethyl ether, ether, bis 2-ethoxyethyl, ethyl diglyme
RRQYJINTUHWNHW-UHFFFAOYSA-N
1-ethoxy-2-(2-ethoxyethoxy)ethane
8179
162.23
98+%

Chemical Identifiers

112-36-7
162.23
RRQYJINTUHWNHW-UHFFFAOYSA-N
8179
1-ethoxy-2-(2-ethoxyethoxy)ethane
C8H18O3
MFCD00009254
1-ethoxy-2-2-ethoxyethoxy ethane, 2-ethoxyethyl ether, 3,6,9-trioxaundecane, bis 2-ethoxyethyl ether, degdee, diethyl carbitol, diethyldiethylene glycol, diethylene glycol diethyl ether, ether, bis 2-ethoxyethyl, ethyl diglyme
CHEBI:44664
CCOCCOCCOCC
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing.

GHS Signal Word: Warning

EINECSNumber : 203-963-7

SDS Documents