Learn More
2-Ethoxyethanol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A16100AP
Description
A widely used solvent for nitrocelluloses and dyes2-Ethoxyethanol is widely used as a varnish remover and degreasing solution. It is a solvent used for nitrocellulose. It is used as a component for dyes, inks, cleaning agents, resins, paints, destaining solution for lipoproteins on polyacrylamide gels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA widely used solvent for nitrocelluloses and dyes2-Ethoxyethanol is widely used as a varnish remover and degreasing solution. It is a solvent used for nitrocellulose. It is used as a component for dyes, inks, cleaning agents, resins, paints, destaining solution for lipoproteins on polyacrylamide gels.
Solubility
Miscible with water, acetone, ethyl acetate, benzene, carbon tetrachloride, diethyl ether, methanol, hydrocarbons and ethanol.
Notes
Incompatible with strong acids, strong bases, strong oxidizing agents, aluminum, alkalies and copper.
Specifications
2-Ethoxyethanol | |
-90°C | |
134°C to 135°C | |
Ether-like | |
1.4085 | |
MFCD00002869 | |
UN1171 | |
14,3750 | |
Miscible with water,acetone,ethyl acetate,benzene,carbon tetrachloride,diethyl ether,methanol,hydrocarbons and ethanol. | |
CCOCCO | |
90.12 | |
CHEBI:46788 | |
99% |
110-80-5 | |
0.931 | |
44°C (111°F) | |
C4H10O2 | |
CH3CH2OCH2CH2OH | |
500 mL | |
1098271 | |
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee | |
ZNQVEEAIQZEUHB-UHFFFAOYSA-N | |
2-ethoxyethan-1-ol | |
8076 | |
90.12 |
Chemical Identifiers
110-80-5 | |
90.12 | |
ZNQVEEAIQZEUHB-UHFFFAOYSA-N | |
8076 | |
2-ethoxyethan-1-ol |
C4H10O2 | |
MFCD00002869 | |
cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee | |
CHEBI:46788 | |
CCOCCO |
Safety and Handling
GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P308+P313-P311-P330-P370+P378q-P501c
H226-H302-H331-H360FD
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOETHYL ETHER
EINECSNumber : 203-804-1
RTECSNumber : KK8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only