Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethoxycyclohexanone, 98%

Chemical Identifiers

Chemical Name or Material 2-Ethoxycyclohexanone
CAS 33371-97-0
Molecular Formula C8H14O2
Formula Weight 142.2
MDL Number MFCD00015377
UN Number UN1224
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2761303

  SDS
alfa aesar™
H2761303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2761306

  SDS
alfa aesar™
H2761306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2761314

  SDS
alfa aesar™
H2761314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.2
Chemical Name or Material 2-Ethoxycyclohexanone
Assay 98%
Flash Point 54°C (129°F)
Boiling Point 85°C (15mmHg)
Molecular Formula C8H14O2
MDL Number MFCD00015377
CAS 33371-97-0
UN Number UN1224
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.